Learn More
Medchemexpress LLC Br-PEG6-C2-acid | 309916-91-4 | 417.29 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY138362
Encompass_Preferred
Br-PEG6-C2-acid is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs are molecules that leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins. This is achieved by connecting two different ligands with a linker, one binding to an E3 ubiquitin ligase and the other to the target protein.
  • PEG-based PROTAC linker
  • For research use only
  • Exploits intracellular ubiquitin-proteasome system
  • Connects E3 ubiquitin ligase ligand and target protein ligand
Catalog No. 50-003-48851