Learn More
Medchemexpress LLC Br-PEG8-OH | 1527515-88-3 | MFCD31656972 | ≥97.0% | 433.33 g·mol⁻¹ | C16H33BrO8 | 500 MG
SDP
Catalog No. 5000281929
Encompass_Preferred
Br-PEG8-OH is a brominated polyethylene glycol (PEG8) derivative with a terminal hydroxyl group, used as a reactive PEG linker in chemical synthesis and bioconjugation. The terminal bromine enables alkylation and coupling reactions to introduce PEG spacers or modify biomolecules. Supplied as a colorless to light yellow liquid with high purity for research use.
  • Terminal bromine enables alkylation and coupling reactions.
  • Eight ethylene glycol units provide a flexible, hydrophilic spacer.
  • Terminal hydroxyl allows further functionalization or conjugation.
  • High supplied purity ≥97.0% suitable for research applications.
  • Physical state liquid, recommended storage at -20°C for long-term stability.
Catalog No. 50-002-81929 Supplier Medchemexpress LLC Supplier No. HYW190743500MG
Br-PEG8-OH is a brominated polyethylene glycol (PEG8) derivative with a terminal hydroxyl group, used as a reactive PEG linker in chemical synthesis and bioconjugation. The terminal bromine enables alkylation and coupling reactions to introduce PEG spacers or modify biomolecules. Supplied as a colorless to light yellow liquid with high purity for research use.
  • Terminal bromine enables alkylation and coupling reactions.
  • Eight ethylene glycol units provide a flexible, hydrophilic spacer.
  • Terminal hydroxyl allows further functionalization or conjugation.
  • High supplied purity ≥97.0% suitable for research applications.
  • Physical state liquid, recommended storage at -20°C for long-term stability.