Learn More
Alkali Scientific Alkali Scientific™ BR1000 BenchBlotter™ 2D Platform Rocker, 115V
SDP
Catalog No. 503029551
Encompass_Preferred
The Alkali Scientific™ BR1000 BenchBlotter is an economical "work-horse" lab rocker, designed for common molecular lab applications such as blotting, staining/destaining, etc. Optimized for Western blot incubations, washing and gel staining Back and forth "teeter - totter" 2D motion Gentle mixing, 12 rpm / +/-8 ° tilt Dual platform accessory doubles work surface Cold room and incubator safe A simple and easy choice for your protein and agarose gels. 
Catalog No. 50-302-9551 Supplier Alkali Scientific Supplier No. RS7007
The Alkali Scientific™ BR1000 BenchBlotter is an economical "work-horse" lab rocker, designed for common molecular lab applications such as blotting, staining/destaining, etc. Optimized for Western blot incubations, washing and gel staining Back and forth "teeter - totter" 2D motion Gentle mixing, 12 rpm / +/-8 ° tilt Dual platform accessory doubles work surface Cold room and incubator safe A simple and easy choice for your protein and agarose gels. 