Learn More
Medchemexpress LLC BR102910 | 2505339-54-6 | 99.8% | 459.30 | C18H14Cl2F2N4O2S | 25 MG
SDP
Catalog No. 5000281540
Encompass_Preferred
BR102910 is a small-molecule selective fibroblast activation protein (FAP) inhibitor intended for research use. It shows potent biochemical activity against FAP and weaker activity against prolyl oligopeptidase (PREP). Supplied at high purity for in vitro and preclinical studies, it is offered in several laboratory-scale package sizes suitable for assay development and compound screening.
  • Selective FAP inhibition (IC50 = 2 nM).
  • Inhibits prolyl oligopeptidase (IC50 = 49.0 μM).
  • High purity suitable for research applications (99.8%).
  • Molecular weight 459.30 and chemical formula C18H14Cl2F2N4O2S.
  • Available in multiple laboratory-scale package sizes, including 25 mg.
Catalog No. 50-002-81540 Supplier Medchemexpress LLC Supplier No. HYW40400225MG
BR102910 is a small-molecule selective fibroblast activation protein (FAP) inhibitor intended for research use. It shows potent biochemical activity against FAP and weaker activity against prolyl oligopeptidase (PREP). Supplied at high purity for in vitro and preclinical studies, it is offered in several laboratory-scale package sizes suitable for assay development and compound screening.
  • Selective FAP inhibition (IC50 = 2 nM).
  • Inhibits prolyl oligopeptidase (IC50 = 49.0 μM).
  • High purity suitable for research applications (99.8%).
  • Molecular weight 459.30 and chemical formula C18H14Cl2F2N4O2S.
  • Available in multiple laboratory-scale package sizes, including 25 mg.