Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Br102910 | 2505339-54-6 | MFCD34565708 | 99.8% | 459.30 | C18H14Cl2F2N4O2S | 100 MG
SDP

Catalog No. 5000281825
Encompass_Preferred

BR102910 is a small-molecule research inhibitor of fibroblast activation protein (FAP) with reported nanomolar potency (IC50 2 nM). It is provided as a high-purity solid for in vitro and preclinical studies and shows additional activity against prolyl oligopeptidase (PREP).

  • High potency FAP inhibitor (IC50 2 nM).
  • Activity against PREP (IC50 49.0 μM) and minimal DPP-4 activity (>100 μM).
  • Purity 99.82%.
  • Molecular weight 459.30; formula C18H14Cl2F2N4O2S.
  • Solid, white to off-white appearance; typical pack size 100 MG.
  • Recommended storage: 4°C for solid; in solvent -80°C (6 months) or -20°C (1 month).
  • Includes structural data (SMILES) for computational and modeling use.

Catalog No. 50-002-81825 Supplier Medchemexpress LLC Supplier No. HYW404002100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

BR102910 is a small-molecule research inhibitor of fibroblast activation protein (FAP) with reported nanomolar potency (IC50 2 nM). It is provided as a high-purity solid for in vitro and preclinical studies and shows additional activity against prolyl oligopeptidase (PREP).

  • High potency FAP inhibitor (IC50 2 nM).
  • Activity against PREP (IC50 49.0 μM) and minimal DPP-4 activity (>100 μM).
  • Purity 99.82%.
  • Molecular weight 459.30; formula C18H14Cl2F2N4O2S.
  • Solid, white to off-white appearance; typical pack size 100 MG.
  • Recommended storage: 4°C for solid; in solvent -80°C (6 months) or -20°C (1 month).
  • Includes structural data (SMILES) for computational and modeling use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.