Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC BR102910 | 2505339-54-6 | MFCD34565708 | 99.8% | 459.30 | C18H14Cl2F2N4O2S | 50 MG
SDP

Catalog No. 5000281455
Encompass_Preferred

A selective fibroblast activation protein (FAP) inhibitor for biochemical and cellular research. It exhibits potent FAP inhibition and weaker activity against prolyl oligopeptidase, making it useful for studies of FAP-related biology and metabolic disease models.

  • Potent FAP inhibition (IC50 = 2 nM).
  • Also inhibits prolyl oligopeptidase (IC50 = 49.0 μM).
  • High purity (99.8%).
  • Solid form, white to off-white powder.
  • Molecular weight 459.30, formula C18H14Cl2F2N4O2S.
  • CAS number 2505339-54-6 for unambiguous identification.
  • Store solid at 4°C; in solvent store at -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-81455 Supplier Medchemexpress LLC Supplier No. HYW40400250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A selective fibroblast activation protein (FAP) inhibitor for biochemical and cellular research. It exhibits potent FAP inhibition and weaker activity against prolyl oligopeptidase, making it useful for studies of FAP-related biology and metabolic disease models.

  • Potent FAP inhibition (IC50 = 2 nM).
  • Also inhibits prolyl oligopeptidase (IC50 = 49.0 μM).
  • High purity (99.8%).
  • Solid form, white to off-white powder.
  • Molecular weight 459.30, formula C18H14Cl2F2N4O2S.
  • CAS number 2505339-54-6 for unambiguous identification.
  • Store solid at 4°C; in solvent store at -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.