Learn More
Hach Company Titration Application Pack Petrochemistry: Bromine Number
Catalog No. 501997523 Shop All Hach Company Products
Encompass
Application Pack for the determination of the Br2/I2 Number in Petrochem. Suited for TitraLab AT1000 Series, Automatic Titrator Models AT1102/AT1112/AT1122/AT1222.
Catalog No. 50-199-7523 Supplier Hach Company Supplier No. AP0018.AT1102
Application Pack for the determination of the Br2/I2 Number in Petrochem. Suited for TitraLab AT1000 Series, Automatic Titrator Models AT1102/AT1112/AT1122/AT1222.