Learn More
Alkali Scientific Alkali Scientific™ BR5000 Everlast Rocker with Variable Speed and Flat Mat - Cold Room Safe
SDP
Catalog No. 503029592
Encompass_Preferred
Everlast Rocker with Variable Speed and Flat Mat Everlastª 5 year warranty Extra-large 16 x 12 in. work surface Stainless-steel platform Dual platform accessory doubles work surface Cold room and incubator safe With an unprecedented 5 year limited warranty, Alkali’s Everlast 247 (BR5000) Rocker is built to last. Twenty four hours a day, seven days a week . . . No problem! The powerful motor and heavy-duty drive system remain reliable and consistent, even under constant usage in a cold room (min. 0°C) or incubator (max. 65°C). The rocking speed and tilt angle can easily be adjusted to vary between gentle and aggressive agitation. Both the standard platform and the (optional) stacking platform are constructed of stainless steel, preventing against any rusting or corrosion. The Everlast 247 is designed to provide years of uninterrupted service.
Catalog No. 50-302-9592 Supplier Alkali Scientific Supplier No. RS7009
Everlast Rocker with Variable Speed and Flat Mat Everlastª 5 year warranty Extra-large 16 x 12 in. work surface Stainless-steel platform Dual platform accessory doubles work surface Cold room and incubator safe With an unprecedented 5 year limited warranty, Alkali’s Everlast 247 (BR5000) Rocker is built to last. Twenty four hours a day, seven days a week . . . No problem! The powerful motor and heavy-duty drive system remain reliable and consistent, even under constant usage in a cold room (min. 0°C) or incubator (max. 65°C). The rocking speed and tilt angle can easily be adjusted to vary between gentle and aggressive agitation. Both the standard platform and the (optional) stacking platform are constructed of stainless steel, preventing against any rusting or corrosion. The Everlast 247 is designed to provide years of uninterrupted service.