Learn More
ATCC Bacillus sp.; Strain: E601
Catalog No. 502381575
Encompass
Bacillus sp. strain E601 is a whole-genome sequence bacterium with applications as an ionizing radiation-sterilization control. This strain is recommended by ATCC for use in the tests described in U.S. Pharmacopeia where only the taxon is specified.
Catalog No. 50-238-1575 Supplier ATCC Supplier No. 27142
Bacillus sp. strain E601 is a whole-genome sequence bacterium with applications as an ionizing radiation-sterilization control. This strain is recommended by ATCC for use in the tests described in U.S. Pharmacopeia where only the taxon is specified.