Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oak Ridge 5 Qt. H-Style Sharps Container

Catalog No. 22730438
Change view
Click to view available options
Capacity (English):
5 qt.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
22-730-438 5 qt.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-730-438 Supplier Oak Ridge Supplier No. 0354110H
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

  • Designed for in unsupervised patient areas
  • Transparent body allows for easy viewing
  • Puncture-resistant, high impact Polypropylene

Specifications

Certifications/Compliance FDA approved, and meet all OSHA and NIOSH regulation
Description Transparent body
Capacity (English) 5 qt.
Color Clear
Dimensions (L x W x H) 23.75 x 17.25 x 11.5 in.
For Use With (Application) Patient room collectors
Material Polypropylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.