Learn More
Braco Manufacturing Premoistened Lens Cleaning Towelettes
Catalog No. 19161914 Shop All Silicone Stirling Paper Products
Encompass
Change view
Click to view available options
Product Type:
Cleaning wipe
Catalog No. Product Type
19-161-914 Cleaning wipe
1 options
Catalog No. 19-161-914 Supplier Braco Manufacturing Supplier No. TW100D
Fast and easy way to keep lenses clean
  • For use on glass, plastic or polycarbonate safety goggles, shields and computer screens
  • Help reduce eyestrain caused by dirty eyewear
  • Excellent cleaning without streaking
  • Silicone-free, antistatic, antifog formula
Order Info
Individually foil wrapped. Packed in ready-to-use dispenser pack with built-in eye hook for easy wall hanging.
Specifications
Description Lens Cleaning Towelettes
Product Type Cleaning wipe
For Use With (Equipment) Lenses
For Use With (Application) For cleaning lenses
Includes Silicone Free formula
Silicone Free Yes
Disposable Single-use