Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bradford Assay Solution (Ready-to-use) [for Protein determination], TCI America™
SDP

Catalog No. B5702500ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B5702500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B5702500ML Supplier TCI America Supplier No. B5702500ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

Synonym Coomassie Brilliant Blue G-250, Acid Blue 90, Brilliant Blue G, CBB G-250

Specifications

Quantity 500 mL
Chemical Name or Material Bradford Assay Solution (Ready-to-use) [for Protein determination]
EINECSNumber (5)-1656
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.