Learn More
Bioworld Bradford Dye Concentrate (5x), 500 mL
Catalog No. 50198424
Encompass_Preferred
Bradford Dye Concentrate, 5x Bradford method utilizes Coomassie Brillant Blue G-250 dye binding to an unknown protein and forming a complex which can be detected spectophotometrically at 595nm. Specifications:Method of choice for protein quantitation. Designed to quantitate 1 to 10µg/ml protein but can be scaled up to quantitate 10 to 100µg/ml simply by increasing the volume of the dye generating a standard curve in the 10 to 100µg/ml range.
Catalog No. 50-198-424 Supplier Bioworld Supplier No. 208300001
Bradford Dye Concentrate, 5x Bradford method utilizes Coomassie Brillant Blue G-250 dye binding to an unknown protein and forming a complex which can be detected spectophotometrically at 595nm. Specifications:Method of choice for protein quantitation. Designed to quantitate 1 to 10µg/ml protein but can be scaled up to quantitate 10 to 100µg/ml simply by increasing the volume of the dye generating a standard curve in the 10 to 100µg/ml range.