Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Bradford Dye Reagent, Ready-to-Use soln.

Catalog No. AAJ61522K2
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ61522K2 1 L
AAJ61522AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ61522K2 Supplier Thermo Scientific Chemicals Supplier No. J61522K2
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

Suitable for protein concentration determination (100 to 1500μg)

The Bradford Reagent is suitable for protein concentration determination (100-1500μg).Bradford dye reagent is used in the determination of protein content in cell fractions and assessing the protein concentrations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bradford Dye Reagent
Color Blue
Name Note Ready-to-Use soln.
UN Number UN1805
Solubility Information Miscible with water.
Quantity 1 L
Physical Form Liquid
At what temperature should I store Bradford Dye Reagent, Ready-to-Use soln.?

The Bradford Dye Reagent, Ready-to-Use soln. should be stored refrigerated at 2-8 degrees C.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.