Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pierce™ Bradford Plus Protein Assay Kits

Catalog No. A55866
Change view
Click to view available options
Concentration:
BSA: 0.125, 0.25, 0.5, 0.75, 1, 1.5, and 2 mg/mL
BSA: 2 mg/mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Concentration
A55866 BSA: 0.125, 0.25, 0.5, 0.75, 1, 1.5, and 2 mg/mL
PI23236 BSA: 2 mg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A55866 Supplier Thermo Scientific™ Supplier No. A55866
Only null left
Add to Cart
Add to Cart

The Pierce Bradford Plus Protein Assay Kit is a ready-to-use, reducing agent-compatible, improved Bradford assay reagent used to quickly measure total protein concentration compared to a protein standard.

The Pierce Bradford Plus Protein Assay Kit is a ready-to-use, reducing agent-compatible, improved Bradford assay reagent used to quickly measure total protein concentration compared to a protein standard. It provides increased linearity and half the protein-to-protein variability of other commercial Bradford assay formulations.

Kits are available with or without Pierce Dilution-Free BSA Protein Standards, which are a set of seven pre-diluted BSA standards, packaged in a multichannel pipette-friendly tubestrip. The tubestrip includes a single empty tube that enables users to add their own sample buffer for the purpose of blank subtraction.

The Pierce Bradford Plus Protein Assay Kit is a ready-to-use, reducing agent-compatible, improved Bradford assay reagent used to quickly measure total protein concentration compared to a protein standard. It provides increased linearity and half the protein-to-protein variability of other commercial Bradford assay formulations.

Kit options include Pierce Dilution-Free BSA Protein Standards, which are a set of seven pre-diluted BSA standards, packaged in a multichannel pipette-friendly tubestrip. The tubestrip includes a single empty tube that enables users to add their own sample buffer for the purpose of blank subtraction.

Features of the Pierce Bradford Plus Protein Assay Kit include:

  • Colorimetric—measure with a standard spectrophotometer or plate reader at 595 nm
  • Easy to use—single reagent; no working reagent preparation required
  • Fast—almost immediate color development; add, mix, and read results
  • Wide assay range—detects protein concentration in the range 1 to 1500 μg/mL
  • Linear—improved linearity and response uniformity compared to traditional Bradford formulations
  • Flexible—microplate and cuvette protocols provided and adaptable to several target working ranges
  • Serial dilution–free—select kit options include Dilution-Free BSA Protein Standards that eliminate the need to perform serial dilutions when generating a standard curve

The assay is performed at room temperature. Simply add the sample to the tube or well containing reagent, and the resultant blue color is measured at 595 nm following a short room-temperature incubation. The protein assay is compatible with most salts, solvents, buffers, thiols, reducing substances, and metal-chelating agents encountered in protein samples. The assay can be performed in either test tube or microplate format.

How the Bradford Plus Assay detects protein

Use of Coomassie G-250 dye in a colorimetric reagent for the detection and quantitation of total protein was first described by Dr. Marion Bradford in 1976. In the acidic environment of the reagent, protein binds to the Coomassie dye. This results in a spectral shift from the reddish/brown form of the dye (absorbance maximum at 465 nm) to the blue form of the dye (absorbance maximum at 610 nm). The difference between the two forms of the dye is greatest at 595 nm, so that is the optimal wavelength to measure the blue color from the Coomassie dye-protein complex. If desired, the blue color can be measured at any wavelength between 575 nm and 615 nm. At the two extremes (575 nm and 615 nm) there is a loss of about 10% in the measured amount of color (absorbance) compared to that obtained at 595 nm.

Development of color in Coomassie dye-based (Bradford) protein assays has been associated with the presence of certain basic amino acids (primarily arginine, lysine, and histidine) in the protein. Van der Waals forces and hydrophobic interactions also participate in the binding of the dye by protein. The number of Coomassie dye ligands bound to each protein molecule is approximately proportional to the number of positive charges found on the protein. Free amino acids, peptides, and low molecular weight proteins do not produce color with Coomassie dye reagents. In general, the mass of a peptide or protein must be at least 3000 Daltons to be assayed with this reagent.

TRUSTED_SUSTAINABILITY

Specifications

Concentration BSA: 0.125, 0.25, 0.5, 0.75, 1, 1.5, and 2 mg/mL
Content And Storage Sufficient for: 630 tube assays or 3160 microplate assays
• Pierce Bradford Plus Assay Reagent, 950 mL
• Dilution-Free BSA Protein Standards (0.125-2 mg/mL), 2 x 1 mL 8-channel tubestrip
Store Bradford Plus Assay Reagent at 4°C.
Assay Bradford Assay
Detection Method Colorimetric
Product Line Pierce
Product Type Protein Quantitation Assay
Specificity Not Target-specific
Sufficient For 630 Tube Assays or 3160 Microplate Assays
For Use With (Application) Solution-based Detection, Absorbance
For Use With (Equipment) Spectrophotometer, Microplate Reader
Kit Contents 2 x 1 mL 8-channel tubestrip, prediluted, dilution-Free BSA protein standards
Quantity 950 mL
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.