Learn More
Apexbio Technology LLC Bradford Protein Assay Kit(Synonyms: Bradford Assay Kit, Bradford Protein Assay, Bradford Protein Quantitation Kit), 500T.
SDP
Catalog No. 5000061164
Encompass_Preferred
Bradford Protein Assay Kit is based on the Bradford method a colorimetric protein quantification approach leveraging Coomassie Brilliant Blue G-250 Under acidic conditions this dye interacts specifically with basic and aromatic amino acid residues notably arginine causing a measurable shift of its absorbance maximum from 465 nm to 595 nm accompanied by a clear solution color change to blue The absorbance intensity at 595 nm directly correlates with protein concentration enabling rapid quantitative analysis Due to its simplicity sensitivity (detection down to 0 5 g) and reliable linearity within 0 1 1 5 mg/mL range it is widely applicable in biomedical protein research
Catalog No. 50-000-61164 Supplier Apexbio Technology LLC Supplier No. K4103500
Bradford Protein Assay Kit is based on the Bradford method a colorimetric protein quantification approach leveraging Coomassie Brilliant Blue G-250 Under acidic conditions this dye interacts specifically with basic and aromatic amino acid residues notably arginine causing a measurable shift of its absorbance maximum from 465 nm to 595 nm accompanied by a clear solution color change to blue The absorbance intensity at 595 nm directly correlates with protein concentration enabling rapid quantitative analysis Due to its simplicity sensitivity (detection down to 0 5 g) and reliable linearity within 0 1 1 5 mg/mL range it is widely applicable in biomedical protein research