Learn More
Crescent Chemical Co Inc BRADFORD REAGENT 5X
SDP
Catalog No. 50248354
Encompass_Preferred
Bradford reagent, 5x Protein dye reagent for protein quantification after Bradford (1). • Precise, reproducible and inexpensive • Fast, only five minutes incubation before reading the sample at 595 nm • Suitable for micro (1- 25 µg protein/ml) and standard (100 – 100
Catalog No. 50-248-354 Supplier Crescent Chemical Co Inc Supplier No. SE39222.03
Bradford reagent, 5x Protein dye reagent for protein quantification after Bradford (1). • Precise, reproducible and inexpensive • Fast, only five minutes incubation before reading the sample at 595 nm • Suitable for micro (1- 25 µg protein/ml) and standard (100 – 100