Learn More
Sigma Aldrich Fine Chemicals Biosciences Bradford Reagent, 500 mL
Catalog No. NC0493460
Encompass
Bradford assay is addition of coomassie brilliant blue G-250 to protein solution. The coomassie blue dye associates with basic and aromatic amino acids, thereby causing shift in absorbance during protein determination.
Catalog No. NC0493460 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. B6916500ML
Bradford assay is addition of coomassie brilliant blue G-250 to protein solution. The coomassie blue dye associates with basic and aromatic amino acids, thereby causing shift in absorbance during protein determination.