Learn More
Bioworld Bradford Reagent, Ready-to-Use, 1 L
Catalog No. 50198427
Encompass_Preferred
Bradford Reagent, Ready to UseBradford method utilizes Coomassie Brillant Blue G-250 dye binding to an unknown protein and forming a complex which can be detected spectophotometrically at 595nm. Method of choice for protein quantitation. Designed to quantitate 1 to 10µg/ml protein but can be scaled up to quantitate 10 to 100µg/ml simply by increasing the volume of the dye generating a standard curve in the 10 to 100µg/ml range. Shipped at ambient temperature, upon receipt store as instructed.
Catalog No. 50-198-427 Supplier Bioworld Supplier No. 208300022
Bradford Reagent, Ready to UseBradford method utilizes Coomassie Brillant Blue G-250 dye binding to an unknown protein and forming a complex which can be detected spectophotometrically at 595nm. Method of choice for protein quantitation. Designed to quantitate 1 to 10µg/ml protein but can be scaled up to quantitate 10 to 100µg/ml simply by increasing the volume of the dye generating a standard curve in the 10 to 100µg/ml range. Shipped at ambient temperature, upon receipt store as instructed.