Learn More
MSC Bradley Combination Shower/Eyewash, Plastic Bowl, Dust Cover, 1 ea
Catalog No. 5000788903
Encompass
This emergency drench shower and eye/face wash unit provides quick and effective eye and face relief. Featuring a plastic bowl, transparent dust cover, and plastic showerhead, it complies with ANSI/ISEA Z358.1 standards. The unit incorporates a self-draining design and galvanized steel piping with a safety yellow coating, ensuring durability and ease of integration into various environments.
  • Effective shower and eye/face washdown coverage
  • Exceeds ANSI/ISEA Z358.1 specifications
  • Self-draining design to reduce standing water
  • Ergonomic hand activation paddle
  • Factory-tested eye/face wash with hinged dust covers
  • Universal identification sign and inspection tag
  • Drench shower flow exceeds 20 GPM at 30 PSI
  • Eye-face wash flow exceeds 3 GPM at 30 PSI
  • Galvanized steel with safety yellow coating
  • Integral strainer reduces debris
Catalog No. 50-007-88903 Supplier MSC Supplier No. 62634399
This emergency drench shower and eye/face wash unit provides quick and effective eye and face relief. Featuring a plastic bowl, transparent dust cover, and plastic showerhead, it complies with ANSI/ISEA Z358.1 standards. The unit incorporates a self-draining design and galvanized steel piping with a safety yellow coating, ensuring durability and ease of integration into various environments.
  • Effective shower and eye/face washdown coverage
  • Exceeds ANSI/ISEA Z358.1 specifications
  • Self-draining design to reduce standing water
  • Ergonomic hand activation paddle
  • Factory-tested eye/face wash with hinged dust covers
  • Universal identification sign and inspection tag
  • Drench shower flow exceeds 20 GPM at 30 PSI
  • Eye-face wash flow exceeds 3 GPM at 30 PSI
  • Galvanized steel with safety yellow coating
  • Integral strainer reduces debris