Learn More
MSC Bradley Corporation Eye/Face Wash, Wall Mount, Dust Cover, 1 ea
Catalog No. 5000789641
Encompass
The Halo Eye-Face Wash unit is designed for emergency eye and face relief. It provides effective washdown protection and features galvanized steel supply and waste pipes with a BRADTECT safety yellow coating. A universal identification sign and inspection tag are included, and the barrier-free design ensures full accessibility.
  • Exceeds minimum water flow of 3 GPM at 30 PSI
  • Barrier-free design for accessibility
  • Galvanized steel supply and waste pipes with BRADTECT safety yellow coating
  • Universal identification sign and inspection tag included
  • UL certified to ANSI/ISEA Z358.1
  • Equipped with a plastic bowl and dust cover
Catalog No. 50-007-89641 Supplier MSC Supplier No. 92031541
The Halo Eye-Face Wash unit is designed for emergency eye and face relief. It provides effective washdown protection and features galvanized steel supply and waste pipes with a BRADTECT safety yellow coating. A universal identification sign and inspection tag are included, and the barrier-free design ensures full accessibility.
  • Exceeds minimum water flow of 3 GPM at 30 PSI
  • Barrier-free design for accessibility
  • Galvanized steel supply and waste pipes with BRADTECT safety yellow coating
  • Universal identification sign and inspection tag included
  • UL certified to ANSI/ISEA Z358.1
  • Equipped with a plastic bowl and dust cover