Learn More
MSC Bradley Drench Shower Eyewash, Galvanized Steel, Plastic Head, 1 ea
Catalog No. 5000793768
Encompass
This drench shower and eyewash station is designed for emergency use, providing immediate decontamination with a drench shower and an eyewash fountain. It features corrosion-resistant components and is activated by a push flag.
  • Bowl, triangular pull rod, and push flag activated
  • Galvanized steel pipe
  • Plastic shower head
  • Bowl flow at 0.4 GPM
  • Corrosion resistant
  • Top or mid supply
Catalog No. 50-007-93768 Supplier MSC Supplier No. 63508279
This drench shower and eyewash station is designed for emergency use, providing immediate decontamination with a drench shower and an eyewash fountain. It features corrosion-resistant components and is activated by a push flag.
  • Bowl, triangular pull rod, and push flag activated
  • Galvanized steel pipe
  • Plastic shower head
  • Bowl flow at 0.4 GPM
  • Corrosion resistant
  • Top or mid supply