Description
- 213.4cm (84 in.) chemically resistant nylon funnel with weighted bottom protects from splashing
- Telescoping aluminum handle extends from 114.3cm (45 in.) to 182.9cm (72 in.)
- Top diameter of 38.1cm (15 in.) and bottom diameter of 12.7cm (5 in.)
- Yellow
Compliance
Compliant with ANSI specifications
Warranty and Services
Full, one-year
Specifications
Specifications
| Includes | Funnel, Handle |
| Product Type | Drench Shower Tester |