Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bradley™ Plastic Eyewash Bowl

Catalog No. 19807530 Shop All Bradley Corporation Products
Change view
Click to view available options
For Use With (Equipment):
Eyewash Station
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
19-807-530 Eyewash Station
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-807-530 Supplier Bradley™ Supplier No. 154058

missing translation for 'validMainframeMsg'

Replacement bowl for emergency eyewash station.

  • Plastic
  • Yellow
  • Fits Bradley Eyewash Stations (109415, 109410, 109417, 109412)

Specifications

Color Yellow
For Use With (Equipment) Eyewash Station
Height (English) 3.87 in.
Height (Metric) 9.8 cm
Material Plastic
Product Type Bowl
Weight (English) 0.65 lb.
Weight (Metric) 0.29 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.