Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bradley™ Eyewash Sign for Emergency Fixtures

Catalog No. 19822022 Shop All Bradley Corporation Products
Change view
Click to view available options
Material:
Plastic
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
19-822-022 Plastic
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-822-022 Supplier Bradley™ Supplier No. 114051

missing translation for 'validMainframeMsg'

Easy to spot visuals of the Bradley Eyewash sign make this easy to spot in the workplace.

  • Dimensions (W x H): 8 x 10.5 in.
  • Yellow plastic

Specifications

For Use With (Equipment) Emergency fixtures
Material Plastic
Quantity 1 Ea.
Height (English) 10.5 in.
Width (English) 8 in.
Displayed Text Eyewash
Product Type Safety Sign
Color Yellow
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.