Learn More
MSC Bradley Eyewash Station, 15 gal, 0.4 GPM, 1 ea
Catalog No. 5000802041
Encompass
The Bradley S19-788 portable eyewash station is designed to provide immediate eye rinsing in emergencies. With a 15-gallon tank and a 0.4 GPM flow rate, it ensures 18 minutes of continuous flow for critical field applications.
  • Pressurized eye wash station
  • Tank capacity: 15 gal
  • Stainless steel construction
  • Maximum flow rate: 0.4 GPM
  • Flow duration: 18 minutes
  • Cart mount location
  • Includes universal identification sign, inspection tag, and protective dust cover
  • Meets ANSI standards
Catalog No. 50-008-02041 Supplier MSC Supplier No. 81226482
The Bradley S19-788 portable eyewash station is designed to provide immediate eye rinsing in emergencies. With a 15-gallon tank and a 0.4 GPM flow rate, it ensures 18 minutes of continuous flow for critical field applications.
  • Pressurized eye wash station
  • Tank capacity: 15 gal
  • Stainless steel construction
  • Maximum flow rate: 0.4 GPM
  • Flow duration: 18 minutes
  • Cart mount location
  • Includes universal identification sign, inspection tag, and protective dust cover
  • Meets ANSI standards