Learn More
MSC Bradley Eyewash Station, Pedestal Mount, Stainless Steel, 1 ea
Catalog No. 5000802607
Encompass
This eyewash station is a pedestal-mount unit designed for emergency eye rinsing. It features a stainless steel bowl and push flag activation, ensuring compliance with ANSI Z358.1 standards for safety equipment.
  • Pedestal mount configuration
  • Stainless steel bowl with cover
  • Deck-mounted installation
  • Activated by push flag mechanism
  • Complies with ANSI Z358.1
  • Overall dimensions: 11 in wide x 40 in high
Catalog No. 50-008-02607 Supplier MSC Supplier No. 43297225
This eyewash station is a pedestal-mount unit designed for emergency eye rinsing. It features a stainless steel bowl and push flag activation, ensuring compliance with ANSI Z358.1 standards for safety equipment.
  • Pedestal mount configuration
  • Stainless steel bowl with cover
  • Deck-mounted installation
  • Activated by push flag mechanism
  • Complies with ANSI Z358.1
  • Overall dimensions: 11 in wide x 40 in high