Learn More
MSC Bradley Halo Drench Shower Eyewash Station, Plastic Showerhead, 1 ea
Catalog No. 5000793470
Encompass
This combined drench shower and eyewash station provides essential emergency washdown protection, designed for immediate and effective rinsing in industrial or laboratory environments. It features a robust construction for reliability and compliance with safety standards.
  • Spintec showerhead ensures consistent water distribution
  • Halo eyewash system offers high-performance rinsing with integral flow control
  • Exceeds ANSI/ISEA Z358.1 specifications for safety equipment
  • Galvanized steel piping protected with a safety yellow coating
  • Equipped with a plastic showerhead, stainless steel shroud, and stainless steel bowl
  • Includes a universal identification sign and inspection tag
  • Self-draining design prevents water accumulation
  • Separate supply and waste pipes meet plumbing requirements
  • Integral strainer reduces debris and prevents clogging
Catalog No. 50-007-93470 Supplier MSC Supplier No. 62634381
This combined drench shower and eyewash station provides essential emergency washdown protection, designed for immediate and effective rinsing in industrial or laboratory environments. It features a robust construction for reliability and compliance with safety standards.
  • Spintec showerhead ensures consistent water distribution
  • Halo eyewash system offers high-performance rinsing with integral flow control
  • Exceeds ANSI/ISEA Z358.1 specifications for safety equipment
  • Galvanized steel piping protected with a safety yellow coating
  • Equipped with a plastic showerhead, stainless steel shroud, and stainless steel bowl
  • Includes a universal identification sign and inspection tag
  • Self-draining design prevents water accumulation
  • Separate supply and waste pipes meet plumbing requirements
  • Integral strainer reduces debris and prevents clogging