Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bradley™ On-Site Gravity-Fed Eyewash Replacement Parts

Catalog No. 19822021 Shop All Bradley Corporation Products
Change view
Click to view available options
Material:
Plastic
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
19-822-021 Plastic
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-822-021 Supplier Bradley™ Supplier No. 136036

missing translation for 'validMainframeMsg'

Replacement part, used with Bradley eyewash units.

  • Cap with hole
  • Plastic

Specifications

Color White
For Use With (Equipment) S19-921, S19-921H, S19-921HR On-Site gravity-fed Eyewash
Includes Hole
Material Plastic
Product Type Cap
Weight (English) 0.02 lb.
Weight (Metric) 0.009 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.