Learn More
MSC Bradley Plumbed Drench Shower, Vertical Mount, Plastic Head, 1 ea
Catalog No. 5000782232
Encompass
The product is a plumbed drench shower system designed for vertical mounting. It features a plastic shower head and is activated by a pull rod.
  • Vertical mount type
  • Plastic shower head material
  • Pull rod activation method
  • Maximum flow rate 22.0 GPM
  • Meets ANSI Z358.1 standards
  • Country of origin: United States
Catalog No. 50-007-82232 Supplier MSC Supplier No. 85605822
The product is a plumbed drench shower system designed for vertical mounting. It features a plastic shower head and is activated by a pull rod.
  • Vertical mount type
  • Plastic shower head material
  • Pull rod activation method
  • Maximum flow rate 22.0 GPM
  • Meets ANSI Z358.1 standards
  • Country of origin: United States