Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bradley Products™ Davidson™ Marking System Dyes
SDP

Catalog No. 23771085
Encompass
Change view
Click to view available options
Quantity:
Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-771085 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dyes are permanent. Never apply these dyes to a living patient. Only use on excised tissue.
Catalog No. 23-771085 Supplier Bradley Products™ Supplier No. 34084
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Dyes are permanent. Never apply these dyes to a living patient. Only use on excised tissue.

Visible, reliable and safe dyes for marking peripheral tumor or surgical margins

  • By applying one dye to superior border, another to inferior border, etc., one can identify the location of tumor or other pathology within a specimen
  • Specimens retain colors and are easily distinguished

Specifications

Capacity 8 oz.
Color Red
Product Type Tissue Marking Dye
Quantity Ea.
For Use With (Application) Use in dying pathological specimens, especially peripheral tumor or surgical margins
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.