Learn More
MSC Bradley Shower and Eye/Face Wash, Barrier-Free, Stainless Steel, 1 ea.
Catalog No. 5000767288
Encompass
The Bradley S19314BF is a barrier-free combination drench shower and Halo eye/face wash unit with a stainless steel bowl, designed for effective emergency washdown. It features a SpinTec showerhead for consistent water distribution and complies with ANSI/ISEA Standard Z358.1.
  • Halo eye/face wash delivers effective washdown coverage and spray pattern
  • Spintec showerhead ensures consistent water distribution
  • Self-draining Aquaduct design
  • Separate supply and waste pipes
  • Ergonomic hand activation eyewash paddle
  • Fully-assembled and factory-tested eye/face wash with hinged dust covers
  • Universal identification sign and inspection tag
  • Galvanized steel piping with safety yellow coating
Catalog No. 50-007-67288 Supplier MSC Supplier No. 63508170
The Bradley S19314BF is a barrier-free combination drench shower and Halo eye/face wash unit with a stainless steel bowl, designed for effective emergency washdown. It features a SpinTec showerhead for consistent water distribution and complies with ANSI/ISEA Standard Z358.1.
  • Halo eye/face wash delivers effective washdown coverage and spray pattern
  • Spintec showerhead ensures consistent water distribution
  • Self-draining Aquaduct design
  • Separate supply and waste pipes
  • Ergonomic hand activation eyewash paddle
  • Fully-assembled and factory-tested eye/face wash with hinged dust covers
  • Universal identification sign and inspection tag
  • Galvanized steel piping with safety yellow coating