Learn More
MSC Bradley Shower Eyewash, Plastic Showerhead, Stainless Steel Bowl, 1 ea
Catalog No. 5000684318
Encompass
This combination drench shower and eyewash unit features a plastic showerhead and a stainless steel bowl, with both hand and foot activated operation. It incorporates a Spintec Showerhead for consistent water distribution, enhancing user washdown effectiveness.
  • Spintec Showerhead ensures consistent water distribution for effective drench shower washdown.
  • Complies with ANSI/ISEA Standard Z358.1.
  • Galvanized steel construction with safety yellow coating.
  • Stainless steel bowl and impact-resistant ABS plastic sprayheads.
  • Features foot pedal or push handle for eyewash activation.
  • Self-draining design for enhanced safety and maintenance.
  • Drench shower flow exceeds 20 GPM at 30 PSI; eyewash flow exceeds 0.4 GPM at 30 PSI.
  • Universal identification sign and inspection tag included.
Catalog No. 50-006-84318 Supplier MSC Supplier No. 63508311
This combination drench shower and eyewash unit features a plastic showerhead and a stainless steel bowl, with both hand and foot activated operation. It incorporates a Spintec Showerhead for consistent water distribution, enhancing user washdown effectiveness.
  • Spintec Showerhead ensures consistent water distribution for effective drench shower washdown.
  • Complies with ANSI/ISEA Standard Z358.1.
  • Galvanized steel construction with safety yellow coating.
  • Stainless steel bowl and impact-resistant ABS plastic sprayheads.
  • Features foot pedal or push handle for eyewash activation.
  • Self-draining design for enhanced safety and maintenance.
  • Drench shower flow exceeds 20 GPM at 30 PSI; eyewash flow exceeds 0.4 GPM at 30 PSI.
  • Universal identification sign and inspection tag included.