Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bradley™ Vertical Supply Drench Shower

Catalog No. 19870006 Shop All Bradley Corporation Products
Change view
Click to view available options
Flow Rate:
20 gpm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Flow Rate
19-870-006 20 gpm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-870-006 Supplier Bradley™ Supplier No. S19130

missing translation for 'validMainframeMsg'

The Bradley Vertical Supply Drench Shower is made from impact-resistant plastic and is bright yellow of high visibility.

Easily mounts in open areas

  • With new SpinTec™ showerhead
  • Galvanized steel protected with Bradtect™ safety yellow coating
  • Universal identification sign and inspection tag included
  • Exceed minimum water flow of 20 gpm at 30psi
  • Full, one-year warranty

Compliance

Complies with ANSI™ Standard Z358.1-2004

Specifications

Activation Pull Rod with Triangular Handle
Certifications/Compliance ANSI Z358.1-2009, EN 15154-1
Flow Rate 20 gpm
For Use With (Application) Unit Design saves Space and Fits easily into any Work Environment
Includes Plastic SpinTec™ shower head, NPT stay open Chrome Plated Brass ball valve, Galvanized Steel, yellow coating pipe and fitting, NPT vertical Water supply, universal identification sign, inspection tag
Material Stainless Steel
Mounting Vertical
Pressure 30 psi
Product Type Drench Shower
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.