Learn More
Brady™ 2-Gallon Spill Response Plus Retail Spill Kit
Catalog No. 19140744 Shop All Brady Corporation Products
Change view
Click to view available options
Type:
Chemical Spill Kit
Spill Kit, Oil-only
Universal Spill Kit
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
19-140-744 Universal Spill Kit
19-140-745 Spill Kit, Oil-only
19-140-746 Chemical Spill Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-140-744 Supplier Brady™ Supplier No. SKASRP
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Includes
2 gal. bucket, 20 pads (7.5 x 10. in. ), 1 disposal bag, tie, 1 pair of nitrile gloves, 1 pair of goggles and instruction sheet
Compact size for convenient handling
  • Highly absorbent, durable and low linting
  • Suitable for hospitality, retail, restaurants or anywhere that requires small pad
  • Absorbs up to 4.5 gal./kit
  • Dia. x H: 24.7 x 26.6cm (9.75 x 10.5 in.)
  • Available types: Universal, oil only and chemical
Specifications
Type Universal Spill Kit
Includes 2 gal. container, 20 pads, 1 bag, tie, pair of gloves, goggles
Dimensions (L x W) 24.7 x 26.6 cm (9.75 x 10.5 in.)