Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ 20 gal. Spill Kit Refill, Reform

Catalog No. 17111232 Shop All Brady Corporation Products
Change view
Click to view available options
Capacity (English):
20 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
17-111-232 20 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-111-232 Supplier Brady™ Supplier No. SKR20R

missing translation for 'validMainframeMsg'

ReForm™ Spill Kit Refill is intended for use with the Reform 20 gal. emergency spill kit. (Refill kit only—container sold separately.)

  • Highly versatile, grey absorbents pick up water-based, petroleum-based and non-aggressive, chemical fluids
  • Improve worker productivity and efficiency by placing spill kits around your facility close to areas prone to spills
  • Spill kits are ideal for fast response to critical accidents

Specifications

Color Dark Green
Capacity (English) 20 gal.
Capacity (Metric) 75.7 L
Type Spill Kit Refill
For Use With (Equipment) Reform 20 gal. emergency spill kit
Includes (12) 15 x 19 in. pads, (3) 3 in. x 12 ft. socks, (2) 18 x 18 in. pillows, 1 pair goggles, 1 pair Nitrile gloves, 3 disposal bags, handbook
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.