Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ 55 gal. Spill Kit Refill, Chemical

Catalog No. 17111237 Shop All Brady Corporation Products
Change view
Click to view available options
Capacity (English):
55 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
17-111-237 55 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-111-237 Supplier Brady™ Supplier No. SKH55R
Add to Cart
Add to Cart

Chemical Spill Kit Refill is intended for use with the Hazwik™ 55 gal. chemical spill kit. (Refill kit only—container sold separately.)

  • Surfactant-treated, brightly colored absorbent picks up a wide range of aggressive chemicals while remaining chemically inert
  • Improve worker productivity and efficiency by placing spill kits around your facility close to areas prone to spills
  • Spill kits are ideal for fast response to critical accidents

Specifications

Color Green
Capacity (English) 55 gal.
Capacity (Metric) 208.2 L
Type Chemical Spill Kit Refill
For Use With (Application) Chemical spills
Includes (50) 15 x 19 in. Pads, (4) 3 in. x 12 ft. socks, (8) 18 x 18 in. pillows, 1 pair goggles, 1 pair Nitrile gloves, 3 disposal bags, handbook
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.