Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ 55 gal. Spill Kit Refill, Oil Only

Catalog No. 17111238 Shop All Brady Corporation Products
Change view
Click to view available options
Capacity (English):
55 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
17-111-238 55 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-111-238 Supplier Brady™ Supplier No. SKO55R
Add to Cart
Add to Cart

Oil-Only Spill Kit Refill is intended for use with the Allwik™ 55 gal. spill kit for oil-based materials. (Refill kit only—container sold separately.)

  • White absorbents pick up petroleum-based fluids, paints and non-water soluble chemicals while repelling water - great for indoor and outdoor use
  • Improve worker productivity and efficiency by placing spill kits around your facility close to areas prone to spills
  • Spill kits are ideal for fast response to critical accidents

Specifications

Color White
Capacity (English) 55 gal.
Capacity (Metric) 208.2 L
Type Spill Kit Refill, Oil-only
For Use With (Application) Oil spills
Includes (50) 15 x 19 in. Pads, (4) 3 in. x 12 ft. socks, (8) 18 x 18 in. pillows, 1 pair goggles, 1 pair Nitrile gloves, 3 disposal bags, handbook
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.