Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Base Neutralizer Spill Kit

Catalog No. 19140749 Shop All Brady Corporation Products
Change view
Click to view available options
Capacity (English):
4.5 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
19-140-749 4.5 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-140-749 Supplier Brady™ Supplier No. SKABKTBASE
  • $317.00
    $210.65 / Each of 1

    Save $106.35

    34% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

6.5 gal. bucket, 2 disposal bags, 1 shaker of base neutralizer (2 lb.), 10 pads (15 x 19 in.), 2 SOCs (3 x 4 in.), 1 pair goggles, 1 pair nitrile gloves, 1 Tyvek suit, 1 small scoop

Specially used to solidify wide range of base spills

  • Polymers will neutralize and change to orange color that indicates neutralization has taken place
  • Absorbs up to 4.5 gal./Kit
  • Dia x H: 35.5 x 45.7cm (14 x 18 in.)
  • Specifications

    Capacity (English) 4.5 gal.
    Material Polyethylene
    Type Base Neutralizing Spill Kit
    Includes (10) 15 x 19 in. pads, (2) 3 in. x 4 ft. socks, 2 disposal bags, 2 lb. Base Neutralizer Shaker, Goggles, Hazmat, Instruction Sheet, Nitrile gloves, Scoop
    Weight (English) 16 lb.
    Weight (Metric) 35.28 kg
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.