Learn More
Brady™ 95 gal. Spill Kit Refill, Chemical
Catalog No. 17111244 Shop All Brady Corporation Products
Change view
Click to view available options
Capacity (English):
95 gal.
Catalog No. Capacity (English)
17-111-244 95 gal.
1 options
Catalog No. 17-111-244 Supplier Brady™ Supplier No. SKH95R
Add to Cart
Add to Cart
Includes
(110) 15 x 19 in. Pads, (12) 3 in. x 4 ft. SOCs, (8) 3 in. x 12 ft. SOCs, (8) 17 x 19 in. Pillows, (1) - Pair Nitrile Gloves, (10) - Disposal Bags, Goggles, Emergency Response Handbook
Chemical Spill Kit Refill is intended for use with the Hazwik™ 95 gal. chemical spill kit. (Refill kit only—container sold separately.)
  • Surfactant-treated, brightly colored absorbent picks up a wide range of aggressive chemicals while remaining chemically inert
  • Improve worker productivity and efficiency by placing spill kits around your facility close to areas prone to spills
  • Spill kits are ideal for fast response to critical accidents
Specifications
Color Green
Capacity (English) 95 gal.
Type Chemical Spill Kit Refill
Includes (110) 15 x 19 in. pads, (8) 3 in. x 12 ft. socks, (12) 3 in. x 4 ft. socks, (8) 18 x 18 in. pillows, 1 pair goggles, 1 pair Nitrile gloves, 3 disposal bags, handbook