Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
Includes
(110) 15 x 19 in. Pads, (12) 3 in. x 4 ft. SOCs, (8) 3 in. x 12 ft. SOCs, (8) 17 x 19 in. Pillows, (1) - Pair Nitrile Gloves, (10) - Disposal Bags, Goggles, Emergency Response Handbook
- Highly versatile, grey absorbents pick up water-based, petroleum-based and non-aggressive, chemical fluids
- Improve worker productivity and efficiency by placing spill kits around your facility close to areas prone to spills
- Spill kits are ideal for fast response to critical accidents
Specifications
Specifications
| Color | Dark Green |
| Capacity (English) | 95 gal. |
| Type | Spill Kit Refill |
| For Use With (Equipment) | Reform 95 gal. emergency spill kit |
| Includes | (110) 15 x 19 in. pads, (12) 3 in. x 4 ft. SOCs, (8) 3 in. x 12 ft. SOCs, (8) 17 x 19 in. pillows, 1 pair Nitrile gloves, 10 disposal bags, goggles, emergency response handbook |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?