Learn More
Brady™ 95 gal. Spill Kit Refill, Universal
Catalog No. 17111247 Shop All Brady Corporation Products
Change view
Click to view available options
Capacity (English):
95 gal.
Catalog No. Capacity (English)
17-111-247 95 gal.
1 options
Catalog No. 17-111-247 Supplier Brady™ Supplier No. SKA95R
  • $738.00
    $574.65 / Each of 1
    Save $163.35
    22% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Includes
(110) 15 x 19 in. Pads, (12) 3 in. x 4 ft. SOCs, (8) 3 in. x 12 ft. SOCs, (8) 17 x 19 in. Pillows, (1) - Pair Nitrile Gloves, (10) - Disposal Bags, Goggles, Emergency Response Handbook
Universal Spill Kit Refill is intended for use with the Allwik™ 95 gal. universal spill kit. (Refill kit only—container sold separately.)
  • Highly versatile, grey absorbents pick up water-based, petroleum-based and non-aggressive, chemical fluids
  • Improve worker productivity and efficiency by placing spill kits around your facility close to areas prone to spills
  • Spill kits are ideal for fast response to critical accidents
Specifications
Color Gray
Capacity (English) 95 gal.
Type Universal Spill Kit Refill