Learn More
MSC Brady Absorbent Roll, Universal, Heavy Weight, Double Perforated, 1 Roll
Catalog No. 5000782116
Encompass
The MRO Plus Double Perforated Absorbent Roll is a robust and low-linting solution designed for various industrial environments. Its heavy-weight construction is ideal for handling continuous leaks and larger spills, effectively reducing material consumption. The 3-ply meltblown polypropylene construction allows for universal absorbency of water-based, petroleum-based, and non-aggressive chemical fluids.
  • Strong, durable, and low linting for industrial applications
  • Heavy-weight design for continuous leaks and larger spills
  • Three-ply meltblown polypropylene construction for enhanced durability
  • Universal absorbency for oil, water, and chemical-based fluids
  • Double perforations enable cost control and waste reduction
  • Dimensions: 30 in W x 150 ft L with a 49 gal absorbency capacity
Catalog No. 50-007-82116 Supplier MSC Supplier No. 00676940
The MRO Plus Double Perforated Absorbent Roll is a robust and low-linting solution designed for various industrial environments. Its heavy-weight construction is ideal for handling continuous leaks and larger spills, effectively reducing material consumption. The 3-ply meltblown polypropylene construction allows for universal absorbency of water-based, petroleum-based, and non-aggressive chemical fluids.
  • Strong, durable, and low linting for industrial applications
  • Heavy-weight design for continuous leaks and larger spills
  • Three-ply meltblown polypropylene construction for enhanced durability
  • Universal absorbency for oil, water, and chemical-based fluids
  • Double perforations enable cost control and waste reduction
  • Dimensions: 30 in W x 150 ft L with a 49 gal absorbency capacity