Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Accessories for BMP™51 and BMP™53 Label Printers

Catalog No. 19000636 Shop All Brady Corporation Products
Change view
Click to view available options
Product Type:
BMP50 Series Spare Battery Tray
Magnet Accessory
Soft Case
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
19-000-636 Magnet Accessory
19-000-632 BMP50 Series Spare Battery Tray
19-000-635 Soft Case
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-000-636 Supplier Brady™ Supplier No. M50MAGNET
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Accessories for BMP™51 and BMP™53 label printers.

  • BMP™51 and BMP™53 Printer Accessories
  • Printer Cases designed for use with BMP™51 and BMP™53 Label Printers from Brady™ protects your printer against impacts and dust while allowing for easy mobility
  • Printer Magnet Attachment allows printer to affix to panel door or metal surface for more hands-free operation or storage
  • Attaches to back of printer with screws to ensure stability
  • Printer Battery Tray fits 8 AA batteries
  • User may pre-load batteries into the spare tray for quick changeover in the field
  • The battery tray snaps in place into the battery compartment

Specifications

Product Type Magnet Accessory
Color Black
For Use With (Equipment) BMP51 Label Printer, BMP53 Label Printer
Sterility Non-sterile
Description BMP50 Series Magnet Accessory
Quantity 1 Ea.
Alternate Part Number M50-MAGNET
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.