Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Accessories for BMP41 Label Printer

Catalog No. 19000209 Shop All Brady Corporation Products
Change view
Click to view available options
Product Type:
Magnetic Attachment
USB Cable
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
19-000-209 Magnetic Attachment
19-000-205 USB Cable
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-000-209 Supplier Brady™ Supplier No. BMP41MAGNET
Add to Cart
Add to Cart

Printer magnet and USB cable for use with BMP41 Label Printers.

  • Printer Magnet Attachment only (printer sold separately), designed for use with the BMP™41 Label Printer from Brady™
  • Allows printer to affix to panel door or metal surface for more hands-free operation or storage
  • Attaches to back of printer with screws to ensure stability
  • Other accessories: Spare USB Cable

Specifications

Product Type Magnetic Attachment
Color Gray
For Use With (Equipment) BMP41 Label Printer
Sterility Non-sterile
Color Capability Gray
Model BMP41-MAGNET
Quantity 1 Ea.
Alternate Part Number BMP41-MAGNET
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.