Learn More
Brady™ Accessories for BMP71 Label Printer
Catalog No. 19000211 Shop All Brady Corporation Products
Change view
Click to view available options
Product Type:
Hard Case
Quick Charger
Rechargeable Battery Pack
Soft Case
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
19-000-211 Quick Charger
19-000-204 Rechargeable Battery Pack
19-000-207 Soft Case
19-000-206 Hard Case
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-000-211 Supplier Brady™ Supplier No. M71QC
Add to Cart
Add to Cart
Printer case, Quick charger and Printer battery for use with BMP-71 Label Printer (printer sold separately).
  • BMP™71 Label Printer Accessories from Brady™
  • Printer Case designed to protect your printer against impacts and dust while allowing for easy mobility.
  • Printer Battery offers long battery life - prints up to 3900 1 in. x 2 in. labels per charge, enables cordless printer operation so you can print labels where you need them, when you need them.
  • BMP™71 Quick Charger, charges battery in one hour.
Specifications
Product Type Quick Charger
For Use With (Equipment) BMP71 Label Printer
Sterility Non-sterile
Quantity 1 Ea.