Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Admittance Signs: DANGER - ELECTRICAL EQUIPMENT - AUTHORIZED PERSONNEL - ONLY

Catalog No. p-169046 Shop All Brady Corporation Products
Change view
Click to view available options
Material:
Fiberglass
Plastic
Polyester
Thickness:
0.006 to 0.01 in.
0.01 in.
0.06 in.
0.1 in.
Width (English):
10 in.
14 in.
Height (English):
7 in.
10 in.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Thickness Width (English) Height (English)
19-102018 Plastic 0.06 in. 14 in. 10 in.
19-016-806 Fiberglass 0.1 in. 10 in. 7 in.
19-035-595 Polyester 0.006 to 0.01 in. 14 in. 10 in.
19-035-750 Polyester 0.01 in. 10 in. 7 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-102018 Supplier Brady™ Supplier No. 22094
Only null left
Add to Cart
Add to Cart

Identify electrical hazard and high voltage areas

  • Available in three materials - plastic, fiberglass and polyester
  • Available in different widths and heights
  • Black/red on white

Specifications

Certifications/Compliance ASTM D 1593, OSHA 1910.145
Material Plastic
Quantity 1 Ea.
Mounting Type Surface Mount
For Use With (Application) Light duty industrial, utility, commercial and institutional environments for both indoor and outdoor use
Height (English) 10 in.
Height (Metric) 25.40 cm
Max. Temperature (English) 175°F
Min. Temperature (English) -40°F
Thickness 0.06 in.
Width (English) 14 in.
Width (Metric) 35.56 cm
Temperature Range (Metric) -40°C to 80°C
Temperature Range (English) -40°F to 175°F
Displayed Text DANGER Electrical Equipment Authorized Personnel Only
Product Type Admittance Sign
Color Black
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.