Learn More
Brady™ Allwik™ 6.5 gal. Spill Bucket, Universal, TAA Compliant
Catalog No. 17111217 Shop All Brady Corporation Products
Change view
Click to view available options
Capacity (English):
9 gal./Bale
Catalog No. Capacity (English)
17-111-217 9 gal./Bale
1 options
Catalog No. 17-111-217 Supplier Brady™ Supplier No. SKABKTTAA
  • $158.30
    $97.65 / Each of 1
    Save $60.65
    38% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Includes
(10) - 15 x 19 in. Pads, (3) - 3 in. x 4 ft. SOCs, (1) - Pair Nitrile Gloves, (2) - Disposal Bags, Instruction Sheet
For fast and small spill response
Intruduction: GSA Schedule Contracts are subject to the Trade Agreements Act (TAA), meaning all products listed on the GSA Schedule Contract be manufactured or 'substantially transformed' in a 'designated country'. The designated countries are composed of:
World Trade Organization Government Procurement Agreement Countries
  • Free Trade Agreement Countries
  • Least Developed Countries; and
  • Caribbean Basin Countries
Specifications
Certifications/Compliance TAA Compliant
Capacity (English) 9 gal./Bale
Material Polyethylene
Type Bucket Spill Kit, Universal
Includes 10 pads, 3 socs (3 x 4 in.), 8 pillows (17 x 19), 1 pair nitrile gloves, 2 Disposal Bags, instruction sheet
Weight (English) 8.5 lb.
Weight (Metric) 18.74 kg