Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Allwik™™ Attack Pac™ Portable Spill Kit

Catalog No. 19000199 Shop All Brady Corporation Products
Change view
Click to view available options
Dimensions (L x W x H):
20 x 16 x 17.5 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W x H)
19-000-199 20 x 16 x 17.5 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-000-199 Supplier Brady™ Supplier No. SKAATK /DEL

missing translation for 'validMainframeMsg'

Includes

15 Pads (15 x 19 in.), 3 Socs (3 in. x 4 ft.), 1 Pair nitrile gloves, 1 Disposable bag, Goggles, Instrution Sheet

Allwik™™ Attack Pac™ Portable Spill Kit is designed for use with spills of any material wherever they occur.

  • Attack Pac Portable Spill Kits combine absorbents, accessories, PPE and instruction guidelines so that you are prepared in an emergency
  • Absorbs up to 7 gallons per kit
  • UV and puncture resistant bag includes a grommet for easy positioning in vehicles and manufacturing work cells
  • Highly versatile, grey absorbents pick up water-based, petroleum-based and non-aggressive, chemical fluids

Specifications

Color Silver
Type Allwik Attack Pac Portable Spill Kit
For Use With (Application) Spill Response
Includes 15 pads, 3 SOCs, 1 pair nitrile gloves, 1 disposal bag, goggles, instruction sheet
Dimensions (L x W x H) 20 x 16 x 17.5 in.
Width (Metric) 40.64 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.