Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ ALLWIK™ Drum Top Cover

Catalog No. 19000157 Shop All Brady Corporation Products
Change view
Click to view available options
For Use With (Equipment):
Drums
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
19-000-157 Drums
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-000-157 Supplier Brady™ Supplier No. DTA25
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ALLWIK™ Drum Top Cover absorbs all liquids.

  • Drum top covers help to maintain clean work surfaces by preventing pump spills and overspray from reaching the floor
  • Highly versatile, grey absorbents pick up water-based, petroleum-based and non-aggressive, chemical fluids
  • Gray color masks spills and stains, helping you resist the urge to pick up the absorbent before it is fully saturated
  • Made of all-purpose polypropylene suitable for most industrial applications

Specifications

Product Type ALLWIK Drum Top Cover
Color Gray
For Use With (Application) Overspray Leaks and Drips, Workstations, Drums - Liquid Storage, Outfalls and Discharge Ponds
For Use With (Equipment) Drums
Material Polypropylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.